Product Details

SNP ID
rs77080639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:44732649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTTTGGGGACAGTGATGAGTAA[A/G]AGTTTTTTGGGACATGGCTGCTTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TSPAN18 PubMed Links

Gene Details

Gene
TSPAN18
Gene Name
tetraspanin 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130783.4 Intron NP_570139.3
XM_005253217.3 Intron XP_005253274.1
XM_006718372.3 Intron XP_006718435.1
XM_006718373.3 Intron XP_006718436.1
XM_011520459.2 Intron XP_011518761.1
XM_017018530.1 Intron XP_016874019.1
XM_017018531.1 Intron XP_016874020.1

View Full Product Details