Product Details

SNP ID
rs75955882
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:45849968 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATTGTTCTGGGTTATGGCCTGG[G/T]CATTAAGGTTTGGGGATTTTTTGTT
Phenotype
MIM: 603732
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CRY2 PubMed Links

Gene Details

Gene
CRY2
Gene Name
cryptochrome circadian clock 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127457.2 Intron NP_001120929.1
NM_021117.3 Intron NP_066940.2

View Full Product Details