Product Details

SNP ID
rs80355554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:45933232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCGGGATAACCACGGAGATGCG[C/T]GACTCACCAAGCAGGGAAGGAAGGA
Phenotype
MIM: 609709 MIM: 608325
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LARGE2 PubMed Links

Gene Details

Gene
LARGE2
Gene Name
LARGE xylosyl- and glucuronyltransferase 2
There are no transcripts associated with this gene.

Gene
PHF21A
Gene Name
PHD finger protein 21A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101802.1 3302 UTR 3 NP_001095272.1
NM_016621.3 3302 UTR 3 NP_057705.3
XM_005252965.4 3302 UTR 3 XP_005253022.1
XM_011520157.2 3302 UTR 3 XP_011518459.1
XM_011520158.2 3302 UTR 3 XP_011518460.1
XM_011520160.2 3302 UTR 3 XP_011518462.1
XM_011520161.2 3302 UTR 3 XP_011518463.1
XM_011520162.2 3302 UTR 3 XP_011518464.1
XM_011520164.2 3302 UTR 3 XP_011518466.1
XM_011520165.2 3302 UTR 3 XP_011518467.1
XM_011520166.2 3302 UTR 3 XP_011518468.1
XM_011520167.2 3302 UTR 3 XP_011518469.1
XM_011520168.2 3302 UTR 3 XP_011518470.1
XM_011520169.2 3302 UTR 3 XP_011518471.1
XM_011520173.2 3302 UTR 3 XP_011518475.1
XM_011520174.2 3302 UTR 3 XP_011518476.1
XM_011520175.2 3302 UTR 3 XP_011518477.1
XM_011520179.2 3302 UTR 3 XP_011518481.1
XM_017017885.1 3302 UTR 3 XP_016873374.1
XM_017017886.1 3302 UTR 3 XP_016873375.1
XM_017017887.1 3302 UTR 3 XP_016873376.1
XM_017017888.1 3302 UTR 3 XP_016873377.1
XM_017017889.1 3302 UTR 3 XP_016873378.1
XM_017017890.1 3302 UTR 3 XP_016873379.1
XM_017017891.1 3302 UTR 3 XP_016873380.1
XM_017017892.1 3302 UTR 3 XP_016873381.1
XM_017017893.1 3302 UTR 3 XP_016873382.1
XM_017017894.1 3302 UTR 3 XP_016873383.1
XM_017017895.1 3302 UTR 3 XP_016873384.1
XM_017017896.1 3302 UTR 3 XP_016873385.1
XM_017017897.1 3302 UTR 3 XP_016873386.1
XM_017017898.1 3302 UTR 3 XP_016873387.1
XM_017017899.1 3302 UTR 3 XP_016873388.1
XM_017017900.1 3302 UTR 3 XP_016873389.1
XM_017017901.1 3302 UTR 3 XP_016873390.1
XM_017017902.1 3302 UTR 3 XP_016873391.1
XM_017017903.1 3302 UTR 3 XP_016873392.1
XM_017017904.1 3302 Intron XP_016873393.1
XM_017017905.1 3302 UTR 3 XP_016873394.1
XM_017017906.1 3302 Intron XP_016873395.1
XM_017017907.1 3302 UTR 3 XP_016873396.1
XM_017017908.1 3302 UTR 3 XP_016873397.1

View Full Product Details