Product Details

SNP ID
rs77561168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47717431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAAAAACTTAAAAACTATGTGTT[A/T]GGAGCCATTTTCCTTCTCTTCAGCC
Phenotype
MIM: 615265
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AGBL2 PubMed Links

Gene Details

Gene
AGBL2
Gene Name
ATP/GTP binding protein like 2
There are no transcripts associated with this gene.

Gene
FNBP4
Gene Name
formin binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318339.1 3235 Silent Mutation CCA,CCT P1017P NP_001305268.1
NM_015308.3 3235 Silent Mutation CCA,CCT P1015P NP_056123.2

View Full Product Details