Product Details

SNP ID
rs80215761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7485943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTTGCAGCTTTTCTGCCCCCGC[C/T]GCAGTGTACCCAGGACCCAGCCATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OLFML1 PubMed Links

Gene Details

Gene
OLFML1
Gene Name
olfactomedin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198474.3 575 Missense Mutation CCG,CTG P23L NP_940876.2
XM_005252878.4 575 Missense Mutation CCG,CTG P23L XP_005252935.1

View Full Product Details