Product Details
- SNP ID
-
rs75979655
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:23532907 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGAAAACAATGATTGTTTTGCTC[A/G]AAATATCAACAGGAAAAACAAAAAT
- Phenotype
-
MIM: 604975
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SOX5
PubMed Links
Gene Details
- Gene
- SOX5
- Gene Name
- SRY-box 5
View Full Product Details