Product Details

SNP ID
rs77584722
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:27081431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGGATTGAGGAGATCTCTGGAG[C/T]GCTTGTCCTTTGGCTGGCGGTTGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C12orf71 PubMed Links

Gene Details

Gene
C12orf71
Gene Name
chromosome 12 open reading frame 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080406.1 1134 Missense Mutation ACT,GCT T185A NP_001073875.1
XM_011520847.2 1134 Missense Mutation ACT,GCT T185A XP_011519149.1

View Full Product Details