Product Details

SNP ID
rs75158113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:27082193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCTCTTGAGTCTGTGTTATCGGA[A/G]CCAATGTCCACGTCCCAGGCCAGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C12orf71 PubMed Links

Gene Details

Gene
C12orf71
Gene Name
chromosome 12 open reading frame 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080406.1 872 Silent Mutation GGC,GGT G97G NP_001073875.1
XM_011520847.2 872 Silent Mutation GGC,GGT G97G XP_011519149.1

View Full Product Details