Product Details

SNP ID
rs79280249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:27696733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGCTGCAAATACTCTGAATGA[C/G]ATCTTCTTAATCCATTTCATCACTT
Phenotype
MIM: 603141 MIM: 610848
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PPFIBP1 PubMed Links

Gene Details

Gene
PPFIBP1
Gene Name
PPFIA binding protein 1
There are no transcripts associated with this gene.

Gene
REP15
Gene Name
RAB15 effector protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029874.1 239 Missense Mutation GAC,GAG D57E NP_001025045.1

View Full Product Details