Product Details

SNP ID
rs76783415
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123265287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAATGATGGCCAGCAGCTCCGC[A/G]TATTTGCTTTGGGGCACCTGGCTGT
Phenotype
MIM: 613541 MIM: 602198
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C12orf65 PubMed Links

Gene Details

Gene
C12orf65
Gene Name
chromosome 12 open reading frame 65
There are no transcripts associated with this gene.

Gene
CDK2AP1
Gene Name
cyclin dependent kinase 2 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270433.1 711 Intron NP_001257362.1
NM_001270434.1 711 Intron NP_001257363.1
NM_004642.3 711 Silent Mutation TAC,TAT Y63Y NP_004633.1

View Full Product Details