Product Details

SNP ID
rs74968466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123409467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGCTTACTGACGTGATTTTGAG[C/T]GTGTTTCCCAGAACTCTGAGCTCTT
Phenotype
MIM: 607240 MIM: 614093
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KMT5A PubMed Links

Gene Details

Gene
KMT5A
Gene Name
lysine methyltransferase 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324504.1 2045 Intron NP_001311433.1
NM_001324505.1 2045 Intron NP_001311434.1
NM_001324506.1 2045 Intron NP_001311435.1
NM_020382.4 2045 Intron NP_065115.3
XM_006719394.3 2045 Intron XP_006719457.1
XM_011538333.2 2045 Intron XP_011536635.1
Gene
RILPL2
Gene Name
Rab interacting lysosomal protein like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145058.2 2045 Intron NP_659495.1
XM_011538012.2 2045 UTR 3 XP_011536314.1
XM_017018959.1 2045 Intron XP_016874448.1

View Full Product Details