Product Details

SNP ID
rs78255092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110017553 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGTTTAAATGACATAAAAGTAC[A/G]TAAAAGAATAGAACTTCAGATTGTT
Phenotype
MIM: 615123
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD13A PubMed Links

Gene Details

Gene
ANKRD13A
Gene Name
ankyrin repeat domain 13A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033121.1 Intron NP_149112.1
XM_005253980.2 Intron XP_005254037.1
XM_005253981.3 Intron XP_005254038.1
XM_005253982.3 Intron XP_005254039.1
XM_005253984.1 Intron XP_005254041.1
XM_011538937.1 Intron XP_011537239.1
XM_011538938.2 Intron XP_011537240.1
XM_017020157.1 Intron XP_016875646.1
XM_017020158.1 Intron XP_016875647.1
XM_017020159.1 Intron XP_016875648.1
XM_017020160.1 Intron XP_016875649.1
XM_017020161.1 Intron XP_016875650.1
XM_017020162.1 Intron XP_016875651.1
XM_017020163.1 Intron XP_016875652.1

View Full Product Details