Product Details

SNP ID
rs75326242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112431976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAATATTTGCAACACATACAGTA[A/G]AGGGTAATTTTCTGGTTATATCAAG
Phenotype
MIM: 176876
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTPN11 PubMed Links

Gene Details

Gene
PTPN11
Gene Name
protein tyrosine phosphatase, non-receptor type 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006719526.2 Intron XP_006719589.1
XM_011538613.2 Intron XP_011536915.1
XM_017019722.1 Intron XP_016875211.1

View Full Product Details