Product Details

SNP ID
rs78482353
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109721681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATTCTAGGGGATCTCTGTGTAT[C/T]CAGGTTGTTCTTAAGCCACAAAAGC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FAM222A PubMed Links

Gene Details

Gene
FAM222A
Gene Name
family with sequence similarity 222 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032829.2 Intron NP_116218.2
XM_006719654.2 Intron XP_006719717.1
XM_017020055.1 Intron XP_016875544.1

View Full Product Details