Product Details

SNP ID
rs74341718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:95020632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAATTTTCAGTTTGTCTCATCAC[A/G]TTAGCAGAGCCTATTCATGCAATTT
Phenotype
MIM: 605250
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCC4 PubMed Links

Gene Details

Gene
ABCC4
Gene Name
ATP binding cassette subfamily C member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105515.2 5113 Intron NP_001098985.1
NM_001301829.1 5113 UTR 3 NP_001288758.1
NM_001301830.1 5113 Intron NP_001288759.1
NM_005845.4 5113 UTR 3 NP_005836.2
XM_005254025.2 5113 UTR 3 XP_005254082.1
XM_017020319.1 5113 UTR 3 XP_016875808.1
XM_017020320.1 5113 Intron XP_016875809.1
XM_017020321.1 5113 UTR 3 XP_016875810.1
XM_017020322.1 5113 Intron XP_016875811.1

View Full Product Details