Product Details

SNP ID
rs78077664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:95815715 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTCAAAATCCAAAACTTAAAAA[A/C]GAGCAAAAATCTTGAATAGGCACTT
Phenotype
MIM: 605898
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UGGT2 PubMed Links

Gene Details

Gene
UGGT2
Gene Name
UDP-glucose glycoprotein glucosyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020121.3 4911 Intron NP_064506.3
XM_011521094.1 4911 UTR 3 XP_011519396.1
XM_011521095.1 4911 UTR 3 XP_011519397.1
XM_011521096.1 4911 UTR 3 XP_011519398.1
XM_011521097.1 4911 Intron XP_011519399.1
XM_011521098.1 4911 UTR 3 XP_011519400.1
XM_011521099.2 4911 UTR 3 XP_011519401.1
XM_011521100.2 4911 Intron XP_011519402.1
XM_011521102.2 4911 UTR 3 XP_011519404.1
XM_011521103.2 4911 UTR 3 XP_011519405.1
XM_017020662.1 4911 UTR 3 XP_016876151.1
XM_017020663.1 4911 UTR 3 XP_016876152.1

View Full Product Details