Product Details

SNP ID
rs76232936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41345327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTTCTTCTCCGTGACTATCAGG[A/G]AGTTGGAAAGTGGGATCCACATTTT
Phenotype
MIM: 612818
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUSAP1 PubMed Links

Gene Details

Gene
NUSAP1
Gene Name
nucleolar and spindle associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243142.1 Intron NP_001230071.1
NM_001243143.1 Intron NP_001230072.1
NM_001243144.1 Intron NP_001230073.1
NM_001301136.1 Intron NP_001288065.1
NM_016359.4 Intron NP_057443.2
NM_018454.7 Intron NP_060924.4
XM_005254428.2 Intron XP_005254485.1
XM_005254430.4 Intron XP_005254487.1
XM_005254431.2 Intron XP_005254488.1
XM_006720559.2 Intron XP_006720622.1
XM_006720560.3 Intron XP_006720623.1
XM_006720561.2 Intron XP_006720624.1
XM_006720562.2 Intron XP_006720625.1
XM_006720563.2 Intron XP_006720626.1
XM_017022294.1 Intron XP_016877783.1
XM_017022295.1 Intron XP_016877784.1

View Full Product Details