Product Details

SNP ID
rs78087533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41984671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCATTAGGAGGAGTGGGAGTGGAG[A/G]AGAAGCACAGAGTTCTTAGCCCCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLA2G4E PubMed Links

Gene Details

Gene
PLA2G4E
Gene Name
phospholipase A2 group IVE
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206670.1 Intron NP_001193599.1
XM_011521237.2 Intron XP_011519539.1
XM_011521238.1 Intron XP_011519540.1
XM_011521239.1 Intron XP_011519541.1
Gene
PLA2G4E-AS1
Gene Name
PLA2G4E antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details