Product Details

SNP ID
rs75424023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43203222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGTGAGTCTAAGGAAGGTGTTCT[C/T]GGGTGGGTTTCGCTCAAAATTGGAG
Phenotype
MIM: 607089 MIM: 177070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCNDBP1 PubMed Links

Gene Details

Gene
CCNDBP1
Gene Name
cyclin D1 binding protein 1
There are no transcripts associated with this gene.

Gene
EPB42
Gene Name
erythrocyte membrane protein band 4.2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000119.2 1704 Missense Mutation AAG,GAG K588E NP_000110.2
NM_001114134.1 1704 Missense Mutation AAG,GAG K558E NP_001107606.1
XM_005254225.1 1704 Missense Mutation AAG,GAG K523E XP_005254282.1
XM_011521349.2 1704 Missense Mutation AAG,GAG K588E XP_011519651.1
XM_011521350.2 1704 Missense Mutation AAG,GAG K588E XP_011519652.1
XM_011521351.2 1704 Missense Mutation AAG,GAG K588E XP_011519653.1
XM_011521352.2 1704 Missense Mutation AAG,GAG K576E XP_011519654.1
XM_011521353.2 1704 Missense Mutation AAG,GAG K553E XP_011519655.1
XM_011521354.2 1704 Missense Mutation AAG,GAG K403E XP_011519656.1

View Full Product Details