Product Details

SNP ID
rs76282442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:77110018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGAAAACCCTAATTCCAGCAAAA[C/T]CAAATGTCAGGTTACTTCCCTGAAC
Phenotype
MIM: 614248
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PEAK1 PubMed Links

Gene Details

Gene
PEAK1
Gene Name
pseudopodium enriched atypical kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024776.3 9609 UTR 3 NP_079052.2
XM_005254670.2 9609 UTR 3 XP_005254727.1
XM_005254671.2 9609 UTR 3 XP_005254728.1
XM_005254673.2 9609 UTR 3 XP_005254730.1
XM_005254674.2 9609 UTR 3 XP_005254731.1
XM_005254675.4 9609 UTR 3 XP_005254732.1
XM_011522033.2 9609 UTR 3 XP_011520335.1
XM_011522034.2 9609 UTR 3 XP_011520336.1
XM_011522035.2 9609 UTR 3 XP_011520337.1
XM_011522036.2 9609 UTR 3 XP_011520338.1
XM_011522037.2 9609 UTR 3 XP_011520339.1
XM_011522038.2 9609 UTR 3 XP_011520340.1
XM_011522039.2 9609 UTR 3 XP_011520341.1
XM_011522040.1 9609 Intron XP_011520342.1
XM_011522041.1 9609 Intron XP_011520343.1
XM_011522042.2 9609 UTR 3 XP_011520344.1

View Full Product Details