Product Details

SNP ID
rs77300664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:101849963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAGAATGAATGACATTTGTCCT[C/T]GAAGAGAAAGAGACTTTATATGTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4F13P PubMed Links

Gene Details

Gene
OR4F13P
Gene Name
olfactory receptor family 4 subfamily F member 13 pseudogene
There are no transcripts associated with this gene.

View Full Product Details