Product Details

SNP ID
rs77094688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:10387672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTATAGTGAGTGTGCACAATCT[A/G]TTTCATTTGATGTAATCAAAGTTTA
Phenotype
MIM: 613645
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATF7IP2 PubMed Links

Gene Details

Gene
ATF7IP2
Gene Name
activating transcription factor 7 interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256160.1 Intron NP_001243089.1
NM_024997.3 Intron NP_079273.2
XM_006720953.3 Intron XP_006721016.1
XM_006720954.3 Intron XP_006721017.1
XM_011522666.2 Intron XP_011520968.1
XM_017023705.1 Intron XP_016879194.1
XM_017023706.1 Intron XP_016879195.1
XM_017023707.1 Intron XP_016879196.1
XM_017023708.1 Intron XP_016879197.1
XM_017023709.1 Intron XP_016879198.1
XM_017023710.1 Intron XP_016879199.1
XM_017023711.1 Intron XP_016879200.1

View Full Product Details