Product Details
- SNP ID
-
rs76162029
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:11281309 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGGCTGAGCCAGCCAACCTGTGAG[C/T]AGGTGGAACTCTGTGGGCTGTGAGT
- Phenotype
-
MIM: 182880
MIM: 182890
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRM1
PubMed Links
Gene Details
- Gene
- PRM1
- Gene Name
- protamine 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002761.2 |
27 |
UTR 5 |
|
|
NP_002752.1 |
- Gene
- PRM2
- Gene Name
- protamine 2
There are no transcripts associated with this gene.
- Gene
- PRM3
- Gene Name
- protamine 3
There are no transcripts associated with this gene.
View Full Product Details