Product Details

SNP ID
rs77529526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:15704608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCGCCTTAGAGGTGAGTTTTCAA[G/T]ATTTGAATTGGAAGAAGGTGAAATG
Phenotype
MIM: 160745 MIM: 609449
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MYH11 PubMed Links

Gene Details

Gene
MYH11
Gene Name
myosin heavy chain 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040113.1 Intron NP_001035202.1
NM_001040114.1 Intron NP_001035203.1
NM_002474.2 Intron NP_002465.1
NM_022844.2 Intron NP_074035.1
XM_011522502.2 Intron XP_011520804.1
XM_017023250.1 Intron XP_016878739.1
Gene
NDE1
Gene Name
nudE neurodevelopment protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143979.1 Intron NP_001137451.1
NM_017668.2 Intron NP_060138.1
XM_005255396.4 Intron XP_005255453.1
XM_006720897.3 Intron XP_006720960.1
XM_006720900.3 Intron XP_006720963.1
XM_011522553.2 Intron XP_011520855.1
XM_017023349.1 Intron XP_016878838.1
XM_017023350.1 Intron XP_016878839.1
XM_017023351.1 Intron XP_016878840.1
XM_017023352.1 Intron XP_016878841.1
XM_017023353.1 Intron XP_016878842.1
XM_017023354.1 Intron XP_016878843.1
XM_017023355.1 Intron XP_016878844.1
XM_017023356.1 Intron XP_016878845.1
XM_017023357.1 Intron XP_016878846.1

View Full Product Details