Product Details

SNP ID
rs76518069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:18792053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACATTTTTTGGCCCTCAGAAGT[A/T]CATTCCTAAAGATTTCAACTACTCT
Phenotype
MIM: 607669 MIM: 603674
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARL6IP1 PubMed Links

Gene Details

Gene
ARL6IP1
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001313858.1 2023 UTR 3 NP_001300787.1
NM_015161.2 2023 UTR 3 NP_055976.1
Gene
RPS15A
Gene Name
ribosomal protein S15a
There are no transcripts associated with this gene.

View Full Product Details