Product Details

SNP ID
rs76715570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:21249988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGGCTGAAGGAGCAGGCTCAGA[A/G]GAATGCCAGGAGGCAGATCAAAGAG
Phenotype
MIM: 609901 MIM: 123740
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKS4B PubMed Links

Gene Details

Gene
ANKS4B
Gene Name
ankyrin repeat and sterile alpha motif domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145865.2 465 Missense Mutation AAG,AGG K141R NP_665872.2
Gene
CRYM
Gene Name
crystallin mu
There are no transcripts associated with this gene.

View Full Product Details