Product Details

SNP ID
rs76243342
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23604098 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTAATCCTCTATCTCAGGCAGTG[C/T]GCACACACATACGGGATCTGTTCCA
Phenotype
MIM: 603836 MIM: 610355
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFAB1 PubMed Links

Gene Details

Gene
NDUFAB1
Gene Name
NADH:ubiquinone oxidoreductase subunit AB1
There are no transcripts associated with this gene.

Gene
PALB2
Gene Name
partner and localizer of BRCA2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024675.3 Intron NP_078951.2
XM_011545946.2 Intron XP_011544248.1
XM_011545947.2 Intron XP_011544249.1
XM_011545948.2 Intron XP_011544250.1
XM_017023671.1 Intron XP_016879160.1
XM_017023672.1 Intron XP_016879161.1
XM_017023673.1 Intron XP_016879162.1

View Full Product Details