Product Details

SNP ID
rs77130343
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:27226762 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGATATTGCAGATCTTCACCGC[A/G]TCGGGGTACGTCTCCCGGATGTATT
Phenotype
MIM: 611917
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KDM8 PubMed Links

Gene Details

Gene
KDM8
Gene Name
lysine demethylase 8
There are no transcripts associated with this gene.

Gene
NSMCE1
Gene Name
NSE1 homolog, SMC5-SMC6 complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145080.3 674 Silent Mutation GAC,GAT D186D NP_659547.2
XM_006721023.3 674 Silent Mutation GAC,GAT D186D XP_006721086.1
XM_011545761.2 674 Silent Mutation GAC,GAT D223D XP_011544063.1
XM_011545762.1 674 Silent Mutation GAC,GAT D90D XP_011544064.1
XM_017023040.1 674 Silent Mutation GAC,GAT D90D XP_016878529.1
XM_017023041.1 674 Silent Mutation GAC,GAT D90D XP_016878530.1
XM_017023042.1 674 Silent Mutation GAC,GAT D90D XP_016878531.1

View Full Product Details