Product Details
- SNP ID
-
rs74907775
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:28260383 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTGTGACCACAGGCTCTGGGAGT[A/G]ATCTGCAAGGGAGACAGGGGTCACC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SBK1
PubMed Links
Gene Details
- Gene
- SBK1
- Gene Name
- SH3 domain binding kinase 1
View Full Product Details