Product Details

SNP ID
rs79199963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30582331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGGTTTTCAGTGAAGGATGGGA[C/T]GTGAACACGGGGCCCTGTGTGCTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF688 PubMed Links

Gene Details

Gene
ZNF688
Gene Name
zinc finger protein 688
There are no transcripts associated with this gene.

Gene
ZNF785
Gene Name
zinc finger protein 785
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152458.6 1913 UTR 3 NP_689671.2
XM_011545752.2 1913 UTR 3 XP_011544054.1
XM_011545753.2 1913 Intron XP_011544055.1
XM_017022967.1 1913 Intron XP_016878456.1

View Full Product Details