Product Details

SNP ID
rs75174484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:636230 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCCACGCTGGGCCGGATCCAGGT[A/C]CTGCCGCAGCACGTGCAAGATGGGA
Phenotype
MIM: 608021
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC100287175 PubMed Links

Gene Details

Gene
LOC100287175
Gene Name
uncharacterized LOC100287175
There are no transcripts associated with this gene.

Gene
MCRIP2
Gene Name
MAPK regulated corepressor interacting protein 2
There are no transcripts associated with this gene.

Gene
METTL26
Gene Name
methyltransferase like 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040160.2 135 Missense Mutation GAC,TAC D21Y NP_001035250.1
NM_001040161.2 135 Missense Mutation GAC,TAC D21Y NP_001035251.1
NM_001040162.2 135 Missense Mutation GAC,TAC D21Y NP_001035252.1
NM_001040165.2 135 Missense Mutation GAC,TAC D21Y NP_001035255.1
NM_001288710.1 135 Missense Mutation GAC,TAC D21Y NP_001275639.1
NM_032366.4 135 Missense Mutation GAC,TAC D21Y NP_115742.3
XM_011522713.2 135 Missense Mutation GAC,TAC D21Y XP_011521015.1
XM_011522714.2 135 Missense Mutation GAC,TAC D21Y XP_011521016.1
Gene
RAB40C
Gene Name
RAB40C, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
WFIKKN1
Gene Name
WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 1
There are no transcripts associated with this gene.

View Full Product Details