Product Details
- SNP ID
-
rs77068987
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:763790 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCATCCCCCAGCATCCCCTCCCCC[G/T]TCCTCCTCTGCTCCACCTCAGATCT
- Phenotype
-
MIM: 601051
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR662
PubMed Links
Gene Details
- Gene
- MIR662
- Gene Name
- microRNA 662
There are no transcripts associated with this gene.
- Gene
- MSLN
- Gene Name
- mesothelin
View Full Product Details