Product Details

SNP ID
rs75674320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56362186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGGGACAAGGAAATGCTGAGAGG[C/G]GAGGGGACCCATGGGGCACACTCAT
Phenotype
MIM: 603243 MIM: 139311
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AMFR PubMed Links

Gene Details

Gene
AMFR
Gene Name
autocrine motility factor receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144.5 2485 UTR 3 NP_001135.3
NM_001323511.1 2485 UTR 3 NP_001310440.1
NM_001323512.1 2485 UTR 3 NP_001310441.1
XM_005255890.3 2485 UTR 3 XP_005255947.1
Gene
GNAO1
Gene Name
G protein subunit alpha o1
There are no transcripts associated with this gene.

View Full Product Details