Product Details

SNP ID
rs74901418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57516419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCTCTGCCACCTTCTCCTGGAA[C/T]TGCTTCTTCAGCTTGCCATGCACAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC102A PubMed Links

Gene Details

Gene
CCDC102A
Gene Name
coiled-coil domain containing 102A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033212.3 1506 Silent Mutation CAA,CAG Q431Q NP_149989.2
XM_011523469.2 1506 Silent Mutation CAA,CAG Q431Q XP_011521771.1

View Full Product Details