Product Details
- SNP ID
-
rs75005894
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:48170166 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCTGCCTCTCCCCCACAGAGAAG[A/T]TTCCACCGTTTTCCACCACATCTGT
- Phenotype
-
MIM: 607040
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ABCC11
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61739606] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABCC11
- Gene Name
- ATP binding cassette subfamily C member 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032583.3 |
4180 |
Missense Mutation |
AAC,ATC |
N1277I |
NP_115972.2 |
NM_033151.3 |
4180 |
Missense Mutation |
AAC,ATC |
N1277I |
NP_149163.2 |
NM_145186.2 |
4180 |
Intron |
|
|
NP_660187.1 |
XM_011523397.1 |
4180 |
Missense Mutation |
AAC,ATC |
N958I |
XP_011521699.1 |
XM_011523398.2 |
4180 |
Missense Mutation |
AAC,ATC |
N654I |
XP_011521700.1 |
XM_017023795.1 |
4180 |
Missense Mutation |
AAC,ATC |
N1277I |
XP_016879284.1 |
XM_017023796.1 |
4180 |
Missense Mutation |
AAC,ATC |
N1277I |
XP_016879285.1 |
XM_017023797.1 |
4180 |
Missense Mutation |
AAC,ATC |
N1277I |
XP_016879286.1 |
XM_017023798.1 |
4180 |
Missense Mutation |
AAC,ATC |
N1277I |
XP_016879287.1 |
XM_017023799.1 |
4180 |
Missense Mutation |
AAC,ATC |
N1277I |
XP_016879288.1 |
XM_017023800.1 |
4180 |
Missense Mutation |
AAC,ATC |
N1277I |
XP_016879289.1 |
XM_017023801.1 |
4180 |
Missense Mutation |
AAC,ATC |
N1241I |
XP_016879290.1 |
XM_017023802.1 |
4180 |
Missense Mutation |
AAC,ATC |
N958I |
XP_016879291.1 |
XM_017023803.1 |
4180 |
Intron |
|
|
XP_016879292.1 |
View Full Product Details