Product Details

SNP ID
rs78203502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:77283803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAATTGAGCTAGAAGCCTACTG[G/T]CAACCTGTCTGTTCCTCAGAGCAGG
Phenotype
MIM: 607512
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADAMTS18 PubMed Links

Gene Details

Gene
ADAMTS18
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001326358.1 3649 UTR 3 NP_001313287.1
NM_199355.3 3649 UTR 3 NP_955387.1
XM_011522923.1 3649 UTR 3 XP_011521225.1
XM_011522924.1 3649 UTR 3 XP_011521226.1
XM_017022988.1 3649 UTR 3 XP_016878477.1
XM_017022989.1 3649 UTR 3 XP_016878478.1

View Full Product Details