Product Details

SNP ID
rs77384806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:87832313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCCTGGCTCAGGATGGTGTCTC[A/G]GGGCTAGGATTATGGTCAGGAGTCC
Phenotype
MIM: 600182
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6775 PubMed Links

Gene Details

Gene
MIR6775
Gene Name
microRNA 6775
There are no transcripts associated with this gene.

Gene
SLC7A5
Gene Name
solute carrier family 7 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006721286.3 1896 UTR 3 XP_006721349.2
XM_017023735.1 1896 Intron XP_016879224.1
XM_017023736.1 1896 Intron XP_016879225.1
XM_017023737.1 1896 Intron XP_016879226.1

View Full Product Details