Product Details

SNP ID
rs75592986
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:17696415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCCCTCAACCCCCCCACCACCC[A/G]TTCATGGGGTTTTTCGCTTTGTGTT
Phenotype
MIM: 607642
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAI1 PubMed Links

Gene Details

Gene
RAI1
Gene Name
retinoic acid induced 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030665.3 Intron NP_109590.3
XM_017024025.1 Intron XP_016879514.1
XM_017024026.1 Intron XP_016879515.1
XM_017024027.1 Intron XP_016879516.1
XM_017024028.1 Intron XP_016879517.1

View Full Product Details