Product Details

SNP ID
rs76619946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2419170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAAATAGGACCGCTGGTTTTTAA[C/T]TTGTGAATATTCTTATTTAAAAACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC284009 PubMed Links

Gene Details

Gene
LOC284009
Gene Name
uncharacterized LOC284009
There are no transcripts associated with this gene.

Gene
METTL16
Gene Name
methyltransferase like 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024086.3 2680 UTR 3 NP_076991.3
XM_005256801.2 2680 UTR 3 XP_005256858.1
XM_017025045.1 2680 UTR 3 XP_016880534.1
XM_017025046.1 2680 UTR 3 XP_016880535.1

View Full Product Details