Product Details

SNP ID
rs77845899
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:21241643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGGCAGGAGCATCTGCCACAG[A/G]TCAAGGGGCAGGGATGTCGGGGGTA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
NATD1 PubMed Links

Gene Details

Gene
NATD1
Gene Name
N-acetyltransferase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152914.2 2014 UTR 3 NP_690878.2

View Full Product Details