Product Details

SNP ID
rs79285623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8377741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCCAAGCTTTAAATAATCACTA[A/C]AACACAAGGGAAAGCCCAACATTCA
Phenotype
MIM: 603704
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KRBA2 PubMed Links

Gene Details

Gene
KRBA2
Gene Name
KRAB-A domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304947.1 Intron NP_001291876.1
NM_213597.3 Intron NP_998762.1
XM_005256443.2 Intron XP_005256500.1
XM_011523643.1 Intron XP_011521945.1
XM_011523644.1 Intron XP_011521946.1
XM_017024163.1 Intron XP_016879652.1
XM_017024164.1 Intron XP_016879653.1
XM_017024165.1 Intron XP_016879654.1
Gene
RPL26
Gene Name
ribosomal protein L26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000987.3 Intron NP_000978.1
NM_001315530.1 Intron NP_001302459.1
NM_001315531.1 Intron NP_001302460.1

View Full Product Details