Product Details

SNP ID
rs77377013
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8392101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGGGAGACCTCTGGCTCCGTTC[C/G]TGGGGCACTCTGCTGGGGTGGCCTC
Phenotype
MIM: 603704
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RNF222 PubMed Links

Gene Details

Gene
RNF222
Gene Name
ring finger protein 222
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146684.2 1755 UTR 3 NP_001140156.1
XM_011523978.2 1755 UTR 3 XP_011522280.1
XM_011523979.2 1755 UTR 3 XP_011522281.1
XM_011523980.2 1755 UTR 3 XP_011522282.1
XM_011523981.2 1755 UTR 3 XP_011522283.1
Gene
RPL26
Gene Name
ribosomal protein L26
There are no transcripts associated with this gene.

View Full Product Details