Product Details

SNP ID
rs79390440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47944487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAAATCCCCAGGAGCACATGGGA[C/T]GGGGTAGCCTGACAGCCCAATAAAG
Phenotype
MIM: 603287
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PNPO PubMed Links

Gene Details

Gene
PNPO
Gene Name
pyridoxamine 5'-phosphate oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018129.3 Intron NP_060599.1
XM_005257500.3 Intron XP_005257557.1
XM_011524968.2 Intron XP_011523270.1
XM_017024813.1 Intron XP_016880302.1
Gene
PRR15L
Gene Name
proline rich 15 like
There are no transcripts associated with this gene.

Gene
SP2-AS1
Gene Name
SP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details