Product Details

SNP ID
rs78549188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50563575 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTCTAGGCTAGCATCTCTTCTC[A/G]CCTGCCCTATCTCCAGGTGCTAGTG
Phenotype
MIM: 604065 MIM: 613939
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CACNA1G PubMed Links

Gene Details

Gene
CACNA1G
Gene Name
calcium voltage-gated channel subunit alpha1 G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256324.1 Intron NP_001243253.1
NM_001256325.1 Intron NP_001243254.1
NM_001256326.1 Intron NP_001243255.1
NM_001256327.1 Intron NP_001243256.1
NM_001256328.1 Intron NP_001243257.1
NM_001256329.1 Intron NP_001243258.1
NM_001256330.1 Intron NP_001243259.1
NM_001256331.1 Intron NP_001243260.1
NM_001256332.1 Intron NP_001243261.1
NM_001256333.1 Intron NP_001243262.1
NM_001256334.1 Intron NP_001243263.1
NM_001256359.1 Intron NP_001243288.1
NM_001256360.1 Intron NP_001243289.1
NM_001256361.1 Intron NP_001243290.1
NM_018896.4 Intron NP_061496.2
NM_198376.2 Intron NP_938190.1
NM_198377.2 Intron NP_938191.2
NM_198378.2 Intron NP_938192.1
NM_198379.2 Intron NP_938193.1
NM_198380.2 Intron NP_938194.1
NM_198382.2 Intron NP_938196.1
NM_198383.2 Intron NP_938197.1
NM_198384.2 Intron NP_938198.1
NM_198385.2 Intron NP_938199.1
NM_198386.2 Intron NP_938200.1
NM_198387.2 Intron NP_938201.1
NM_198388.2 Intron NP_938202.1
NM_198396.2 Intron NP_938406.1
XM_006722160.3 Intron XP_006722223.1
XM_006722161.3 Intron XP_006722224.1
Gene
CACNA1G-AS1
Gene Name
CACNA1G antisense RNA 1
There are no transcripts associated with this gene.

Gene
SPATA20
Gene Name
spermatogenesis associated 20
There are no transcripts associated with this gene.

View Full Product Details