Product Details

SNP ID
rs77321952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:56985394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCATAGGGCGGTCCAGGCGGTT[C/T]TAGCACTGGATTTGGAAACTTTGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCPEP1 PubMed Links

Gene Details

Gene
SCPEP1
Gene Name
serine carboxypeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021626.2 277 Missense Mutation TCT,TTT S81F NP_067639.1
XM_005257557.2 277 Missense Mutation TCT,TTT S81F XP_005257614.1

View Full Product Details