Product Details

SNP ID
rs79081307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38877957 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGCCCCCAGCTCACCTGGGATGA[C/T]TCCATGTTCTCCCCACAGCCATGGG
Phenotype
MIM: 602920
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LASP1 PubMed Links

Gene Details

Gene
LASP1
Gene Name
LIM and SH3 protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271608.1 Intron NP_001258537.1
NM_006148.3 Intron NP_006139.1

View Full Product Details