Product Details

SNP ID
rs74893159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75792900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATAGAGAGGGCTTCAAGCGCATG[A/G]CCTATTGCCAAGGCTCAGCAACAGT
Phenotype
MIM: 616375
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4738 PubMed Links

Gene Details

Gene
MIR4738
Gene Name
microRNA 4738
There are no transcripts associated with this gene.

Gene
UNK
Gene Name
unkempt family zinc finger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080419.2 Intron NP_001073888.2
XM_006722145.3 Intron XP_006722208.1
XM_006722148.2 Intron XP_006722211.1
XM_011525400.1 Intron XP_011523702.1
XM_011525401.2 Intron XP_011523703.1
XM_017025247.1 Intron XP_016880736.1
XM_017025248.1 Intron XP_016880737.1
XM_017025249.1 Intron XP_016880738.1
XM_017025250.1 Intron XP_016880739.1

View Full Product Details