Product Details

SNP ID
rs75690512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78424502 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTAAAGTTCCCTGATTCTTAAT[A/G]TGTAATGTCTGCCCTATGTGTACAT
Phenotype
MIM: 610063 MIM: 614942
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAH17 PubMed Links

Gene Details

Gene
DNAH17
Gene Name
dynein axonemal heavy chain 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173628.3 6452 Intron NP_775899.3
XM_011525416.2 6452 Intron XP_011523718.1
XM_017025261.1 6452 Intron XP_016880750.1
Gene
PGS1
Gene Name
phosphatidylglycerophosphate synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024419.4 6452 UTR 3 NP_077733.3
XM_011525487.2 6452 UTR 3 XP_011523789.1
XM_011525488.2 6452 UTR 3 XP_011523790.1
XM_011525489.2 6452 UTR 3 XP_011523791.1
XM_017025357.1 6452 UTR 3 XP_016880846.1
XM_017025358.1 6452 UTR 3 XP_016880847.1
XM_017025359.1 6452 UTR 3 XP_016880848.1
XM_017025360.1 6452 UTR 3 XP_016880849.1
XM_017025361.1 6452 UTR 3 XP_016880850.1
XM_017025362.1 6452 UTR 3 XP_016880851.1
XM_017025363.1 6452 UTR 3 XP_016880852.1
XM_017025364.1 6452 UTR 3 XP_016880853.1
XM_017025365.1 6452 UTR 3 XP_016880854.1
XM_017025366.1 6452 UTR 3 XP_016880855.1

View Full Product Details