Product Details
- SNP ID
-
rs79588023
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40766277 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTTAATGAAATGTCAAAAAAGGC[C/T]CCCAAAATAAAAACAAAAATTAGGG
- Phenotype
-
MIM: 616675
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT26
PubMed Links
Gene Details
- Gene
- KRT26
- Gene Name
- keratin 26
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181539.4 |
1691 |
UTR 3 |
|
|
NP_853517.2 |
View Full Product Details