Product Details

SNP ID
rs75100886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:80138487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTATCCTACTTAACATTTCACAA[C/T]GTGCACCACATGGAATACGATTCTA
Phenotype
MIM: 608546
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF4A3 PubMed Links

Gene Details

Gene
EIF4A3
Gene Name
eukaryotic translation initiation factor 4A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014740.3 Intron NP_055555.1
XM_011525522.1 Intron XP_011523824.1

View Full Product Details